Assay Design



Gene Transcripts

Gene Symbol
Lin28b
Entrez Gene ID
380669
Gene Name
lin-28 homolog B (C. elegans)
Gene Aliases
2810403D23Rik, D030047M17Rik, Lin-28.2
Chromosome Location
Chr.10: 45376619 - 45487303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031772.2 NP_001026942.1 4 - 5 585 63
GenBank mRNA BC089037.1 - 4 - 5 576 63
DQ127225.1 - 4 - 5 585 63

View Full Product Details