Assay Design
Gene Transcripts
- Gene Symbol
- Hus1
- Entrez Gene ID
-
15574
- Gene Name
- Hus1 homolog (S. pombe)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 8993137 - 9011534 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
Y16894.2 |
- |
8 - 9 |
963 |
90 |
View Full Product Details