Assay Design



Gene Transcripts

Gene Symbol
Kremen1
Entrez Gene ID
84035
Gene Name
kringle containing transmembrane protein 1
Gene Aliases
AV002070, Kremen, Krm1
Chromosome Location
Chr.11: 5191551 - 5261636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032396.3 NP_115772.2 6 - 7 1050 72
GenBank mRNA AB059617.1 - 6 - 7 1013 72
AK141321.1 - 6 - 7 1062 72
BC049771.1 - 6 - 7 974 72
BC082546.1 - 6 - 7 1050 72
BC138464.1 - 6 - 7 1058 72
BC138465.1 - 6 - 7 1058 72

View Full Product Details