Assay Design



Gene Transcripts

Gene Symbol
Edn2
Entrez Gene ID
13615
Gene Name
endothelin 2
Gene Aliases
VIC
Chromosome Location
Chr.4: 120161424 - 120167360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007902.2 NP_031928.2 4 - 5 519 99
GenBank mRNA BC037042.1 - 4 - 5 748 99

View Full Product Details