Assay Design
Gene Transcripts
- Gene Symbol
- Pabpc4
- Entrez Gene ID
-
230721
- Gene Name
- poly(A) binding protein, cytoplasmic 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 123262387 - 123298832 on Build GRCm38
- Gene Symbol
- Gm5088
- Entrez Gene ID
-
328451
- Gene Name
- poly(A)-binding protein, cytoplasmic pseudogene
- Gene Aliases
-
EG328451, Pabpc4l
- Chromosome Location
-
Chr.14: 89896224 - 89899447 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002862.3 |
|
1 - 1 |
|
84 |
GenBank mRNA |
AK048750.1 |
- |
1 - 1 |
|
84 |
View Full Product Details