Assay Design



Gene Transcripts

Gene Symbol
Ppie
Entrez Gene ID
56031
Gene Name
peptidylprolyl isomerase E (cyclophilin E)
Gene Aliases
2010010D16Rik, Cyp33
Chromosome Location
Chr.4: 123127114 - 123140000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019489.5 NP_062362.1 6 - 7 389 67
GenBank mRNA AK008177.1 - 6 - 7 390 67
AK042165.1 - 3 - 4 2973 67
BC045154.1 - 6 - 7 384 67

View Full Product Details