Assay Design



Gene Transcripts

Gene Symbol
Il20ra
Entrez Gene ID
237313
Gene Name
interleukin 20 receptor, alpha
Gene Aliases
E230031K19
Chromosome Location
Chr.10: 19711673 - 19760053 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172786.2 NP_766374.1 2 - 3 277 72
GenBank mRNA AK054215.1 - 2 - 3 278 72
BC056628.1 - 2 - 3 261 72

View Full Product Details