Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029868.2 | NP_001025039.1 | 6 - 7 | 1298 | 89 |
NM_001290540.1 | NP_001277469.1 | 4 - 5 | 953 | 89 | |
GenBank mRNA | AK046101.1 | - | 4 - 5 | 955 | 89 |
BC056462.1 | - | 6 - 7 | 644 | 89 |