Assay Design



Gene Transcripts

Gene Symbol
Kdm2b
Entrez Gene ID
30841
Gene Name
lysine (K)-specific demethylase 2B
Gene Aliases
Cxxc2, Fbl10, Fbxl10, Jhdm1b, PCCX2
Chromosome Location
Chr.5: 122870662 - 122989127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK043352.1 - 10 - 11 1279 80
AK087953.1 - 10 - 11 1085 80
AK129479.1 - 10 - 11 1072 80
BC065090.1 - 10 - 11 1271 80
BC068239.1 - 7 - 8 825 80

View Full Product Details