Assay Design



Gene Transcripts

Gene Symbol
Mtss1
Entrez Gene ID
211401
Gene Name
metastasis suppressor 1
Gene Aliases
2310003N14Rik, BC024131, D130001D01Rik, Mim, mKIAA0429
Chromosome Location
Chr.15: 58941234 - 59082325 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144800.2 NP_659049.2 6 - 7 965 77
GenBank mRNA AK030533.1 - 4 - 5 359 77
AY214918.1 - 6 - 7 466 77
BC024131.1 - 6 - 7 930 77

View Full Product Details