Assay Design



Gene Transcripts

Gene Symbol
Myrf
Entrez Gene ID
225908
Gene Name
myelin regulatory factor
Gene Aliases
6030439E18, Gm1804, Gm98, Mrf
Chromosome Location
Chr.19: 10208271 - 10240781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033481.1 NP_001028653.1 25 - 26 3403 67
GenBank mRNA AK039649.1 - 16 - 17 1765 67
BC157942.1 - 23 - 24 2875 67

View Full Product Details