Assay Design



Gene Transcripts

Gene Symbol
Myrf
Entrez Gene ID
225908
Gene Name
myelin regulatory factor
Gene Aliases
6030439E18, Gm1804, Gm98, Mrf
Chromosome Location
Chr.19: 10208271 - 10240781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033481.1 NP_001028653.1 6 - 7 1134 80
GenBank mRNA AK141695.1 - 6 - 7 1135 80
BC157942.1 - 3 - 4 525 80

View Full Product Details