Assay Design



Gene Transcripts

Gene Symbol
Micall2
Entrez Gene ID
231830
Gene Name
MICAL-like 2
Gene Aliases
A930021H16Rik, JRAB, MICAL-L2, mFLJ00139
Chromosome Location
Chr.5: 139706692 - 139736333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174850.3 NP_777275.2 6 - 7 1952 69
GenBank mRNA AB182579.1 - 6 - 7 1803 69
AK044547.1 - 6 - 7 1804 69
AK085948.1 - 6 - 7 1923 69
AK151086.1 - 6 - 7 1951 69
AK164043.1 - 6 - 7 1923 69
AK165558.1 - 6 - 7 1802 69
AK220308.1 - 6 - 7 1729 69
BC043300.1 - 6 - 7 1766 69

View Full Product Details