Assay Design



Gene Transcripts

Gene Symbol
Wbscr16
Entrez Gene ID
94254
Gene Name
Williams-Beuren syndrome chromosome region 16 homolog (human)
Gene Aliases
5730496C04Rik, AU019812
Chromosome Location
Chr.5: 134148058 - 134177301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033572.2 NP_291050.1 2 - 3 500 88
GenBank mRNA AF410456.1 - 2 - 3 479 88
AK005040.1 - 2 - 3 446 88
AK017733.1 - 2 - 3 499 88
BC024714.1 - 2 - 3 472 88
BC055335.1 - 2 - 3 498 88

View Full Product Details