Assay Design



Gene Transcripts

Gene Symbol
Wbscr16
Entrez Gene ID
94254
Gene Name
Williams-Beuren syndrome chromosome region 16 homolog (human)
Gene Aliases
5730496C04Rik, AU019812
Chromosome Location
Chr.5: 134148058 - 134177301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033572.2 NP_291050.1 5 - 6 743 96
GenBank mRNA AF410456.1 - 5 - 6 722 96
AK005040.1 - 5 - 6 689 96
AK017733.1 - 5 - 6 742 96
BC024714.1 - 5 - 6 715 96
BC055335.1 - 5 - 6 741 96

View Full Product Details