Assay Design



Gene Transcripts

Gene Symbol
Tecpr1
Entrez Gene ID
70381
Gene Name
tectonin beta-propeller repeat containing 1
Gene Aliases
2210010N04Rik, mKIAA1358
Chromosome Location
Chr.5: 144194442 - 144223608 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027410.1 NP_081686.1 9 - 10 1275 51
GenBank mRNA AB093295.1 - 8 - 9 1444 51
AK032858.1 - 9 - 10 1249 51
AK034020.1 - 8 - 9 1853 51
AK169888.1 - 9 - 10 1222 51
AK170108.1 - 9 - 10 1222 51
AK186825.1 - 1 - 2 35 51
AK188059.1 - 1 - 2 35 51
AK193764.1 - 1 - 2 35 51
AK218769.1 - 1 - 2 35 51
BC046968.1 - 9 - 10 1275 51
BC052185.1 - 3 - 4 1852 51

View Full Product Details