Assay Design



Gene Transcripts

Gene Symbol
Dnajc30
Entrez Gene ID
66114
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 30
Gene Aliases
1300007M11Rik, Wbscr18
Chromosome Location
Chr.5: 135064206 - 135065365 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025362.3 NP_079638.2 1 - 1 59 67
GenBank mRNA AF412026.1 - 1 - 1 62 67
AK019256.1 - 1 - 1 38 67

View Full Product Details