Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007583.2 | NP_031609.1 | 2 - 3 | 689 | 68 |
GenBank mRNA | AF077739.1 | - | 2 - 3 | 689 | 68 |
AK045362.1 | - | - | 1438 | 68 | |
BC103563.1 | - | 2 - 3 | 370 | 68 | |
BC103564.1 | - | 2 - 3 | 369 | 68 | |
BC103664.1 | - | 2 - 3 | 369 | 68 |