Assay Design



Gene Transcripts

Gene Symbol
Ncf4
Entrez Gene ID
17972
Gene Name
neutrophil cytosolic factor 4
Gene Aliases
AI451400, p40phox
Chromosome Location
Chr.15: 78244164 - 78262582 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008677.2 NP_032703.2 6 - 7 771 95
GenBank mRNA AB002665.1 - 6 - 7 748 95
AK150510.1 - 6 - 7 753 95
AK170854.1 - 3 - 4 279 95
AK171315.1 - 6 - 7 549 95
BC025517.1 - 6 - 7 600 95
FJ168470.1 - 6 - 7 562 95
U59488.1 - 6 - 7 718 95

View Full Product Details