Assay Design



Gene Transcripts

Gene Symbol
Oosp3
Entrez Gene ID
225923
Gene Name
oocyte secreted protein 3
Gene Aliases
Gm97
Chromosome Location
Chr.19: 11697055 - 11711858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033283.2 NP_001028455.2 2 - 3 298 109
GenBank mRNA AK143410.1 - 2 - 3 299 109
BC094934.1 - 2 - 3 260 109
BC141228.1 - 2 - 3 275 109

View Full Product Details