Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013774.2 | NP_001013796.2 | 8 - 9 | 1124 | 67 |
GenBank mRNA | AK136027.1 | - | 4 - 5 | 642 | 67 |
AY950703.1 | - | 8 - 9 | 1125 | 67 | |
FJ717332.1 | - | 6 - 7 | 891 | 67 | |
FJ717333.1 | - | 7 - 8 | 954 | 67 |