Assay Design



Gene Transcripts

Gene Symbol
Ide
Entrez Gene ID
15925
Gene Name
insulin degrading enzyme
Gene Aliases
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location
Chr.19: 37268741 - 37334544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031156.3 NP_112419.3 3 - 4 690 62
GenBank mRNA AJ278422.1 - 3 - 4 710 62
AK004972.2 - 3 - 4 690 62
AK014703.1 - - 610 62
AK128986.1 - 3 - 4 707 62
BC041675.1 - 3 - 4 722 62

View Full Product Details