Assay Design



Gene Transcripts

Gene Symbol
Mapk10
Entrez Gene ID
26414
Gene Name
mitogen-activated protein kinase 10
Gene Aliases
C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK
Chromosome Location
Chr.5: 102907948 - 103211339 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009158.2 NP_033184.2 13 - 14 1812 138
GenBank mRNA AB005665.1 - 10 - 11 1195 138
L35236.1 - 13 - 14 138

View Full Product Details