Assay Design



Gene Transcripts

Gene Symbol
Brdt
Entrez Gene ID
114642
Gene Name
bromodomain, testis-specific
Gene Aliases
7420412D09Rik, Brd6, Fsrg3
Chromosome Location
Chr.5: 107331091 - 107387061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054054.2 NP_473395.2 13 - 14 2389 124
GenBank mRNA AK039465.1 - 1 - 2 155 121
BY724250.1 - 1 - 2 155 121

View Full Product Details