Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033561.1 | NP_291039.1 | 4 - 5 | 435 | 88 |
GenBank mRNA | AK045004.1 | - | 4 - 5 | 437 | 88 |
AK088818.1 | - | 4 - 5 | 447 | 88 | |
AK132034.1 | - | 4 - 5 | 455 | 88 | |
AK135390.1 | - | 4 - 5 | 436 | 88 | |
AK149841.1 | - | 4 - 5 | 421 | 88 | |
AK150088.1 | - | 4 - 5 | 444 | 88 | |
AK166567.1 | - | 4 - 5 | 436 | 88 | |
AK169312.1 | - | 4 - 5 | 418 | 88 | |
AK169353.1 | - | 4 - 5 | 436 | 88 | |
AK182256.1 | - | 1 - 2 | 82 | 88 | |
AK184202.1 | - | 1 - 2 | 82 | 88 | |
AK184510.1 | - | 1 - 2 | 82 | 88 | |
AK192457.1 | - | 1 - 2 | 82 | 88 | |
AK193051.1 | - | 1 - 2 | 82 | 88 | |
AK193739.1 | - | 1 - 2 | 88 | ||
AK196800.1 | - | 1 - 2 | 82 | 88 | |
AK197761.1 | - | 1 - 2 | 82 | 88 | |
AK200533.1 | - | 1 - 2 | 82 | 88 |