Assay Design
Gene Transcripts
- Gene Symbol
- Fbrsl1
- Entrez Gene ID
-
381668
- Gene Name
- fibrosin-like 1
- Gene Aliases
-
2410025L10Rik, AI467568, Gm1049, mKIAA1545
- Chromosome Location
-
Chr.5: 110361751 - 110449233 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028596.2 |
NP_082872.2 |
3 - 4 |
774 |
124 |
GenBank mRNA |
BC151196.1 |
- |
4 - 5 |
655 |
124 |
View Full Product Details