Assay Design
Gene Transcripts
- Gene Symbol
- Galnt9
- Entrez Gene ID
-
231605
- Gene Name
- UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9
- Gene Aliases
-
GalNAc-T9
- Chromosome Location
-
Chr.5: 110544350 - 110621383 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198306.3 |
NP_938048.2 |
2 - 3 |
775 |
77 |
GenBank mRNA |
AK032568.1 |
- |
2 - 3 |
777 |
77 |
View Full Product Details