Assay Design



Gene Transcripts

Gene Symbol
Myo1h
Entrez Gene ID
231646
Gene Name
myosin 1H
Gene Aliases
4631401O15Rik
Chromosome Location
Chr.5: 114288899 - 114364576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164573.1 NP_001158045.1 11 - 12 85

View Full Product Details