Assay Design



Gene Transcripts

Gene Symbol
Sfswap
Entrez Gene ID
231769
Gene Name
splicing factor, suppressor of white-apricot homolog (Drosophila)
Gene Aliases
1190005N23Rik, 6330437E22Rik, AI197402, AW212079, SWAP, Sfrs8, Srsf8
Chromosome Location
Chr.5: 129501231 - 129571384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172276.3 NP_758480.2 9 - 10 1590 70
GenBank mRNA AK031872.1 - 9 - 10 1581 70
AK088182.1 - 9 - 10 1534 70
AK140369.1 - 9 - 10 1583 70
BC023268.1 - 4 - 5 621 70
BC057314.1 - 9 - 10 3501 70

View Full Product Details