Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163575.1 | NP_001157047.1 | 3 - 4 | 527 | 63 |
NM_001163576.1 | NP_001157048.1 | 2 - 3 | 631 | 63 | |
GenBank mRNA | AK028373.1 | - | 3 - 4 | 475 | 63 |
AK035309.1 | - | 3 - 4 | 521 | 63 | |
AK046755.1 | - | 2 - 3 | 612 | 63 | |
AK146336.1 | - | 3 - 4 | 528 | 63 | |
AK153205.1 | - | 2 - 3 | 368 | 63 | |
AK166291.1 | - | 3 - 4 | 478 | 63 | |
BC024074.1 | - | 3 - 4 | 484 | 63 | |
BY742887.1 | - | 3 - 4 | 528 | 63 |