Assay Design



Gene Transcripts

Gene Symbol
Ms4a8a
Entrez Gene ID
64381
Gene Name
membrane-spanning 4-domains, subfamily A, member 8A
Gene Aliases
2010004L09Rik, AI481240, CD20L5, Ms4a8
Chromosome Location
Chr.19: 11067471 - 11081103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022430.2 NP_071875.2 6 - 7 781 66
GenBank mRNA AB026044.1 - 6 - 7 731 66
AF237906.1 - 6 - 7 719 66
AK008099.1 - 6 - 7 720 66
AY039038.1 - 6 - 7 719 66
BC042719.1 - 6 - 7 767 66

View Full Product Details