Assay Design
Gene Transcripts
- Gene Symbol
- Sumf2
- Entrez Gene ID
-
67902
- Gene Name
- sulfatase modifying factor 2
- Gene Aliases
-
2610040F05Rik
- Chromosome Location
-
Chr.5: 129846990 - 129863951 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK166216.1 |
- |
8 - 9 |
829 |
74 |
View Full Product Details