Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289544.1 | NP_001276473.1 | 2 - 3 | 570 | 79 |
NM_028227.3 | NP_082503.2 | 2 - 3 | 442 | 79 | |
GenBank mRNA | AF321920.1 | - | 2 - 3 | 334 | 79 |
AF321921.1 | - | 2 - 3 | 571 | 79 | |
AK190663.1 | - | 1 - 2 | 80 | 79 | |
BC062932.1 | - | 2 - 3 | 419 | 79 | |
BF467152.1 | - | 2 - 3 | 355 | 79 | |
CD355617.1 | - | 2 - 3 | 419 | 79 |