Assay Design



Gene Transcripts

Gene Symbol
Hpse2
Entrez Gene ID
545291
Gene Name
heparanase 2
Gene Aliases
Gm968, Hpa2
Chromosome Location
Chr.19: 42786541 - 43388569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081257.2 NP_001074726.1 10 - 11 1516 108
GenBank mRNA BC158131.1 - 10 - 11 1516 108
BC158133.1 - 10 - 11 1516 108
BC158136.1 - 10 - 11 1516 108

View Full Product Details