Assay Design



Gene Transcripts

Gene Symbol
Tmed11
Entrez Gene ID
67366
Gene Name
transmembrane emp24 protein transport domain containing
Gene Aliases
1810008K16Rik
Chromosome Location
Chr.5: 108777235 - 108795363 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026109.2 NP_080385.2 2 - 3 275 122
GenBank mRNA AK007381.1 - 2 - 3 284 122
AK019032.1 - 2 - 3 264 122
BC132664.1 - 2 - 3 247 122
BC132666.1 - 2 - 3 247 122
BY707479.1 - - 280 122

View Full Product Details