Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253877.1 | NP_001240806.1 | 12 - 13 | 1810 | 93 |
NM_001253878.1 | NP_001240807.1 | 12 - 13 | 1529 | 93 | |
NM_001253879.1 | NP_001240808.1 | 11 - 12 | 1452 | 93 | |
NM_001253880.1 | NP_001240809.1 | 9 - 10 | 1167 | 93 | |
NM_013827.3 | NP_038855.2 | 11 - 12 | 1448 | 93 | |
NR_045608.1 | 13 - 14 | 1516 | 93 | ||
GenBank mRNA | AK030752.1 | - | 11 - 12 | 1398 | 93 |
AK031409.1 | - | 13 - 14 | 1517 | 93 | |
AK032798.1 | - | 12 - 13 | 1771 | 93 | |
AK032819.1 | - | 12 - 13 | 1772 | 93 | |
AK137539.1 | - | 13 - 14 | 1757 | 93 | |
AK139685.1 | - | 12 - 13 | 1530 | 93 | |
BC050826.1 | - | 11 - 12 | 1402 | 93 | |
BC092237.1 | - | 11 - 12 | 1380 | 93 | |
BC100340.1 | - | 9 - 10 | 1098 | 93 | |
S78454.1 | - | 8 - 9 | 852 | 93 |