Assay Design
Gene Transcripts
- Gene Symbol
- Cux2
- Entrez Gene ID
-
13048
- Gene Name
- cut-like homeobox 2
- Gene Aliases
-
1700051K22Rik, Cutl2, Cux-2, ENSMUSG00000072641
- Chromosome Location
-
Chr.5: 121857962 - 122050562 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_007804.2 |
NP_031830.2 |
11 - 12 |
1123 |
66 |
GenBank mRNA |
BC065113.1 |
- |
11 - 12 |
1123 |
66 |
U45665.1 |
- |
11 - 12 |
944 |
66 |
View Full Product Details