Assay Design



Gene Transcripts

Gene Symbol
Bud31
Entrez Gene ID
231889
Gene Name
BUD31 homolog (yeast)
Gene Aliases
C77604, EDG-2, EDG2, G10
Chromosome Location
Chr.5: 145140338 - 145148074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008705.1 NP_001008705.1 4 - 5 467 85
GenBank mRNA BC006861.1 - 5 - 6 501 85
BC057025.1 - 4 - 5 467 85

View Full Product Details