Assay Design



Gene Transcripts

Gene Symbol
Enpp2
Entrez Gene ID
18606
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases
ATX, Npps2, PD-Ialpha, Pdnp2, lysoPLD
Chromosome Location
Chr.15: 54838679 - 54952887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136077.2 NP_001129549.1 17 - 18 1790 89
NM_001285994.1 NP_001272923.1 16 - 17 1634 89
NM_001285995.1 NP_001272924.1 17 - 18 1790 89
NM_015744.3 NP_056559.2 16 - 17 1634 89
GenBank mRNA AF123542.1 - 16 - 17 1490 89
AK088491.1 - 17 - 18 1662 89
AK161144.1 - 16 - 17 1507 89
AK163881.1 - 16 - 17 1508 89
BC003264.1 - 16 - 17 1511 89
BC058759.1 - 16 - 17 1597 89
EU131009.1 - 17 - 18 1605 89
EU131010.1 - 16 - 17 1449 89

View Full Product Details