Assay Design



Gene Transcripts

Gene Symbol
Enpp2
Entrez Gene ID
18606
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases
ATX, Npps2, PD-Ialpha, Pdnp2, lysoPLD
Chromosome Location
Chr.15: 54838679 - 54952887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136077.2 NP_001129549.1 21 - 22 2253 139
NM_001285994.1 NP_001272923.1 21 - 22 2172 139
NM_001285995.1 NP_001272924.1 21 - 22 2241 139
NM_015744.3 NP_056559.2 20 - 21 2097 139
GenBank mRNA AF123542.1 - 20 - 21 1953 139
AK088491.1 - 21 - 22 2113 139
AK161144.1 - 20 - 21 1970 139
AK163881.1 - 20 - 21 1971 139
BC003264.1 - 20 - 21 1974 139
BC058759.1 - 20 - 21 2060 139
EU131009.1 - 21 - 22 2068 139
EU131010.1 - 21 - 22 1987 139

View Full Product Details