Assay Design



Gene Transcripts

Gene Symbol
Rbpms
Entrez Gene ID
19663
Gene Name
RNA binding protein gene with multiple splicing
Gene Aliases
2010300K22Rik, 2700019M19Rik, AU017537, RBP-MS, hermes
Chromosome Location
Chr.8: 33782644 - 33929895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042674.2 NP_001036139.1 6 - 7 851 100
GenBank mRNA AK041118.1 - 6 - 7 795 100

View Full Product Details