Assay Design
Gene Transcripts
- Gene Symbol
- Mboat4
- Entrez Gene ID
-
234155
- Gene Name
- membrane bound O-acyltransferase domain containing 4
- Gene Aliases
-
GOAT, Gm171
- Chromosome Location
-
Chr.8: 34108219 - 34125185 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC147844.1 |
- |
1 - 2 |
169 |
111 |
BC147849.1 |
- |
1 - 2 |
168 |
111 |
View Full Product Details