Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164039.1 | NP_001157511.1 | 2 - 3 | 442 | 96 |
NM_008529.3 | NP_032555.1 | 1 - 2 | 653 | 96 | |
GenBank mRNA | BC002116.1 | - | 1 - 2 | 56 | 96 |
BC019113.1 | - | 1 - 2 | 82 | 96 | |
BC037143.1 | - | 1 - 2 | 67 | 96 |