Assay Design



Gene Transcripts

Gene Symbol
Trappc9
Entrez Gene ID
76510
Gene Name
trafficking protein particle complex 9
Gene Aliases
1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882
Chromosome Location
Chr.15: 72589620 - 73061243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164641.1 NP_001158113.1 2 - 3 835 76
NM_001164642.1 NP_001158114.1 2 - 3 835 76
NM_001164643.1 NP_001158115.1 2 - 3 835 76
NM_180662.2 NP_850993.2 2 - 3 835 76
GenBank mRNA AK028504.1 - 2 - 3 677 76
AK029106.1 - 2 - 3 677 76
AK031788.1 - 2 - 3 683 76
AK138682.1 - 2 - 3 682 76
AK156739.1 - 2 - 3 660 76
BC070463.1 - 2 - 3 835 76

View Full Product Details