Assay Design



Gene Transcripts

Gene Symbol
Hsf1
Entrez Gene ID
15499
Gene Name
heat shock factor 1
Gene Aliases
AA960185, Hsf1alpha, Hsf1beta
Chromosome Location
Chr.15: 76477395 - 76500978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008296.2 NP_032322.1 5 - 6 717 59
GenBank mRNA BC013716.1 - 5 - 6 717 59
BC094064.1 - 5 - 6 727 59
KM043052.1 - 5 - 6 717 59
KM043053.1 - 5 - 6 717 59
KM043054.1 - 5 - 6 717 59
KM043055.1 - 5 - 6 717 59
X61753.1 - 5 - 6 700 59

View Full Product Details