Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025573.3 | NP_079849.1 | 3 - 4 | 659 | 74 |
NR_036616.1 | 4 - 5 | 749 | 74 | ||
GenBank mRNA | AK011621.1 | - | 3 - 4 | 619 | 74 |
AK020084.1 | - | 3 - 4 | 431 | 74 | |
AK159190.1 | - | 3 - 4 | 657 | 74 | |
BC012217.1 | - | 3 - 4 | 659 | 74 | |
CV558546.1 | - | 3 - 4 | 408 | 74 |