Assay Design



Gene Transcripts

Gene Symbol
Cyth3
Entrez Gene ID
19159
Gene Name
cytohesin 3
Gene Aliases
AI648983, ARNO3, CLM3, Grp1, Pscd3
Chromosome Location
Chr.5: 143622447 - 143710250 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163548.1 NP_001157020.1 1 - 2 553 86
GenBank mRNA AK016645.1 - 1 - 2 555 86

View Full Product Details