Assay Design



Gene Transcripts

Gene Symbol
Dtx2
Entrez Gene ID
74198
Gene Name
deltex 2 homolog (Drosophila)
Gene Aliases
2610524D08Rik, AA408415, AU022494, Deltex2
Chromosome Location
Chr.5: 135994800 - 136032881 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256097.1 NP_001243026.1 2 - 3 245 71
GenBank mRNA AK044105.1 - 2 - 3 202 71

View Full Product Details