Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144847.1 | NP_659096.1 | 6 - 7 | 582 | 69 |
GenBank mRNA | BC011468.1 | - | 6 - 7 | 515 | 69 |
BC012437.1 | - | 6 - 7 | 582 | 69 | |
BC025042.1 | - | 1 - 2 | 44 | 69 | |
BC068117.1 | - | 11 - 12 | 1230 | 69 |