Assay Design
Gene Transcripts
- Gene Symbol
- Nrbp2
- Entrez Gene ID
-
223649
- Gene Name
- nuclear receptor binding protein 2
- Gene Aliases
-
BC011468
- Chromosome Location
-
Chr.15: 76085589 - 76091594 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK158425.1 |
- |
4 - 5 |
781 |
75 |
BC068117.1 |
- |
4 - 5 |
642 |
75 |
View Full Product Details